Jenway 3510 Standard Digital pH Meter Only, ATC; 120 VAC

Representative image only.

Jenway 3510 Standard Digital pH Meter Only, ATC; 120 VAC

Jenway | Item: 3545903

Compact unit provides accurate and reliable readings

₹100,000.00INR / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹100,000.00INR / Each

Buy It Together

Current Item

Jenway 3510 Standard Digital pH Meter Only, ATC; 120 VAC
Jenway 3510 Standard Digital pH Meter Only, ATC; 120 VAC
3545903

₹100,000.00INR / Each
+
Jenway 037 701 Thermal printer with IrDA
9995980

₹63,000.00INR / Each
+ + + + +
Jenway 4510 Conductivity/TDS Meter and Probe Stand; 120 VAC
3544002

₹106,000.00INR / Each
=
Total Price 487200.00 ADD 7 ITEMS TO CART

Customers Also Bought

Product Description

Includes

Power cord with US plug.

Key Features

  • Supports one, two, or three point pH calibration
  • Up to three decimal place resolution for accurate readings
  • Stores up to 32 readings
  • 120 VAC operation
  • Comes with US plug
  • 3 year warranty

More About this Item

Benchtop pH meters feature up to three decimal place resolution, support up to three pH calibration points, and offer manual and automatic buffer recognition for DIN, JIS, and NIST buffers. A large LCD displays status and results of pH and temperature readings. Easily export data to a computer or printer via an RS-232 or an analog interface.

Advanced digital pH meters help ensure compliance with Good Laboratory Practice (GLP) guidelines with calibration reminder interval, alarm output, and security code setup features. Both standard and advanced pH meters are available as meter only or as a kit that includes a pH electrode, ATC probe, and buffers.

3 Year WarrantyConformite Europeenne
Specifications
  • Meter TypeBenchtop
  • Display typeBacklit custom LCD
  • ConnectionsBNC
  • Min pH (pH)-2
  • Max pH (pH)19.999
  • pH Resolution0.001, 0.01, 0.1
  • pH Accuracy±0.003
  • Buffer RecognitionDIN, NIST, JIS, and custom
  • CalibrationUser selectable 1, 2 or 3 point
  • Min mV (mV)-1999.9
  • Max mV (mV)1999.9
  • mV Resolution0.1, 1
  • mV Accuracy±0.2
  • Min Temperature (° C)-10
  • Max Temperature (° C)105
  • Min Temperature (° F)14
  • Max Temperature (° F)221
  • Temperature Resolution1°F (0.1°C)
  • Temperature Accuracy±1°F (±0.5°C)
  • Temperature compensationAutomatic or manual from 32 to 212⁰F (0 to 100⁰C)
  • InterfaceAnalog and RS-232
  • OutputAnalog and RS-232
  • Data Logging (points)32 data sets
  • Memory Size32
  • Power (VAC)120
  • Length (cm)25
  • Width (cm)21
  • Height (cm)5.5
  • Width (in)8 1/4
  • Length (in)9 7/8
  • Height (in)2 3/16
  • DescriptionStandard Digital pH Meter Only, ATC; 120 VAC
  • Warranty3 year
Reviews
Q&A