Jenway 354 201 Bench Conductivity/pH Meter Kit, 230 VAC, EU Power Supply

Representative image only.

Jenway 354 201 Bench Conductivity/pH Meter Kit, 230 VAC, EU Power Supply

Jenway | Item: 7900058

Ideal for use in all laboratories where pH and conductivity analyses are required

₹173,000.00INR / Each

₹173,000.00INR / Each

Buy It Together

Current Item

Jenway 354 201 Bench Conductivity/pH Meter Kit, 230 VAC, EU Power Supply
Jenway 354 201 Bench Conductivity/pH Meter Kit, 230 VAC, EU Power Supply
7900058

₹173,000.00INR / Each
+ +
Jenway 3510 Standard Digital pH Meter Only, ATC; 230 VAC/UK
3545901

₹100,000.00INR / Each
+ + =
Total Price 326900.00 ADD 5 ITEMS TO CART

Customers Also Bought

Product Description

Includes

3540 conductivity/ pH meter supplied with glass combination pH electrode (924 005), glass conductivity probe (K= 1 027 013), electrode stand and holder, ATC probe (027 500), BNC shorting plug, pH buffers and EU power supply

Key Features

  • Simultaneous display of pH and conductivity
  • 2 independent channels to prevent interference between probes
  • Additional modes for resistivity, salinity, TDS and mV
  • Data logger with auto save/print options
  • Storage of up to 500 readings (250 for each mode)

More About this Item

The setup menu gives quick and easy access to the whole range of instrument, pH, and conductivity measurement options. View pH/temperature and conductivity/temperature readings at the same time on the LCD display. Calibration of both channels is automatic, with the option to select 1, 2 or 3 calibration points.

Meter ensures compliance with Good Laboratory Practice (GLP) guidelines with calibration reminder interval, operator and sample ID, and security code setup features. Send data to a computer or printer via RS-232 or analog outputs. RS-232 output includes data for both channels as displayed on the screen. The analog output can be switched between the pH and conductivity measurement. Set alarm for each channel with high and low set points.

Specifications
  • Meter TypepH/Conductivity
  • Display type1/8 VGA LCD, backlit
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.001, 0.01, or 0.1 selectable
  • pH Accuracy±0.003 pH
  • Buffer RecognitionDIN, NIST, and JIS
  • Calibration1, 2, or 3 points
  • Min mV (mV)-1999.9
  • Max mV (mV)1999.99
  • mV Resolution0.1 mV
  • mV Accuracy±0.2 mV
  • Min Conductivity0.01 µS
  • Max Conductivity1.999 S
  • Conductivity Resolution0.01 µS to 1 mS
  • Conductivity Accuracy±0.5% ±2 digits
  • Min Temperature (° C)-10
  • Max Temperature (° C)105
  • Min Temperature (° F)14
  • Max Temperature (° F)221
  • Temperature Resolution0.1°
  • Temperature Accuracy±0.5°C (±1°F)
  • Temperature compensationYes
  • Temperature Coefficient0 to 4%/°C
  • InterfaceRS-232
  • OutputRS-232, analog, and alarm outputs
  • Memory Size250 readings
  • Power (VAC)230
  • Length (cm)25
  • Width (cm)21
  • Height (cm)5.5
  • Width (in)8 1/4
  • Length (in)9 7/8
  • Height (in)2 3/16
  • Probe IncludedYes
  • Accessories Included3540 conductivity/ pH meter supplied with glass combination pH electrode (924 005), glass conductivity probe (K= 1 027 013), electrode stand and holder, ATC probe (027 500), BNC shorting plug, pH buffers and EU power supply
  • DescriptionBench Conductivity/pH Meter Kit, 230 VAC, EU Power Supply
Reviews
Q&A