Jenway Benchtop pH Meters
Compact unit provides accurate and reliable readings
  • Supports one, two, or three point pH calibration
  • Up to three decimal place resolution for accurate readings
Benchtop pH meters feature up to three decimal place resolution, support up to three pH calibration points, and offer manual and automatic buffer recognition for DIN, JIS, and NIST buffers. A large LCD displays status and results of pH and temperature readings. Easily export data to a computer or printer via an RS-232 or an analog interface.
Advanced digital pH meters help ensure compliance with Good Laboratory Practice (GLP) guidelines with calibration reminder interval, alarm output, and security code setup features. Both standard and advanced pH meters are available as meter only or as a kit that includes a pH electrode, ATC probe, and buffers.
77,900.00 - ₹180,000.00INR / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Description
Availability Pricing
COMPARE
230 Standard Digital pH Meter Only, ATC; 230 VAC/UK
₹80,800.00INR / Each
COMPARE
230 Standard Digital pH Meter Kit; 230 VAC/UK
₹80,300.00INR / Each
COMPARE
120 Standard Digital pH Meter Only, ATC; 120 VAC
₹77,900.00INR / Each
COMPARE
120 Standard Digital pH Meter Kit; 120 VAC
₹91,900.00INR / Each
COMPARE
230 Standard Digital pH Meter Only, ATC; 230 VAC/EU
₹77,900.00INR / Each
COMPARE
230 Standard Digital pH Meter Kit; 230 VAC/EU
₹82,000.00INR / Each
COMPARE
230 Advanced Digital pH Meter Only with GLP, ATC; 230 VAC/UK
₹115,000.00INR / Each
COMPARE
230 Advanced Digital pH Meter Kit with GLP; 230 VAC/UK
₹131,000.00INR / Each
COMPARE
120 Advanced Digital pH Meter Only with GLP, ATC; 120 VAC
₹180,000.00INR / Each
COMPARE
120 Advanced Digital pH Meter Kit with GLP; 120 VAC
₹111,000.00INR / Each
COMPARE
230 Advanced Digital pH Meter Only with GLP, ATC; 230 VAC/EU
₹178,000.00INR / Each
COMPARE
230 Advanced Digital pH Meter Kit with GLP; 230 VAC/EU
₹131,000.00INR / Each
Showing 1- 12 of 12