Jenway® Bench Combined pH and Conductivity Meters
Ideal for use in all laboratories where pH and conductivity analyses are required
  • Simultaneous display of pH and conductivity
  • 2 independent channels to prevent interference between probes
  • Additional modes for resistivity, salinity, TDS and mV
  • Data logger with auto save/print options
  • Storage of up to 500 readings (250 for each mode)
The setup menu gives quick and easy access to the whole range of instrument, pH, and conductivity measurement options. View pH/temperature and conductivity/temperature readings at the same time on the LCD display. Calibration of both channels is automatic, with the option to select 1, 2 or 3 calibration points.
Meter ensures compliance with Good Laboratory Practice (GLP) guidelines with calibration reminder interval, operator and sample ID, and security code setup features. Send data to a computer or printer via RS-232 or analog outputs. RS-232 output includes data for both channels as displayed on the screen. The analog output can be switched between the pH and conductivity measurement. Set alarm for each channel with high and low set points.
181,000.00 - ₹218,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Bench Conductivity/pH Meter Kit, 230 VAC, EU Power Supply
₹201,000.00INR / Each
COMPARE
Bench Conductivity/pH Meter Kit, 230 VAC, UK Power Supply
₹181,000.00INR / Each
COMPARE
Bench Conductivity/pH Meter Kit, 120 VAC, US Power Supply
₹218,000.00INR / Each