Jenway® Conductivity Cells
Cells for conductivity measurement
  • For use with benchtop and portable meters
Compatible with Jenway meters.
15,300.00 - ₹69,200.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cell Constant Use With
Availability Pricing
COMPARE
K = 0.1 ± 20% Jenway 470 meter
₹15,300.00INR / Each
COMPARE
K = 10 ± 20% Jenway 470 meter
₹17,300.00INR / Each
COMPARE
K = 0.1 ± 20% Jenway meters 4510, 4520 and 3540
₹38,200.00INR / Each
COMPARE
K = 0.1 ± 20% Jenway meters 4510, 4520 and 3540
₹47,100.00INR / Each
COMPARE
K = 1 ± 20% Jenway meters 4510, 4520 and 3540
₹45,500.00INR / Each
COMPARE
K = 10 ± 20% Jenway meters 4510, 4520 and 3540
₹23,200.00INR / Each
COMPARE
K = 1 ± 20% Jenway 470 meter
₹45,300.00INR / Each
COMPARE
Nominal 0.08 Jenway meters 4510, 4520 and 3540
₹69,200.00INR / Each
COMPARE
K = 1 ± 10% Jenway meters 4510, 4520 and 3540
₹33,400.00INR / Each
Showing 1- 9 of 9