Jenway 924 030 Semi-Micro, 90 mm Reach, Glass Bodied Tris Buffer pH Electrode for Biological Buffers, Blood, and Protein Samples

Representative image only.

Jenway 924 030 Semi-Micro, 90 mm Reach, Glass Bodied Tris Buffer pH Electrode for Biological Buffers, Blood, and Protein Samples

Jenway | Item: 9996039

A variety of pH electrodes for your Jenway meters

₹32,700.00INR / Each

₹32,700.00INR / Each

Buy It Together

Current Item

Jenway 924 030 Semi-Micro, 90 mm Reach, Glass Bodied Tris Buffer pH Electrode for Biological Buffers, Blood, and Protein Samples
Jenway 924 030 Semi-Micro, 90 mm Reach, Glass Bodied Tris Buffer pH Electrode for Biological Buffers, Blood, and Protein Samples
9996039

₹32,700.00INR / Each
+ + +
Jenway 3510 Standard Digital pH Meter Only, ATC; 230 VAC/UK
3545901

₹100,000.00INR / Each
+ +
Jenway 025 157 Redox Standard, 200 mV; 500 mL
7900029

₹16,200.00INR / Each
+
Jenway 3510 pH Meter with FREE Electrode, 120 VAC
3545913

₹244,000.00INR / Each
+
Jenway 025 160 pH Electrode Fill Solution, 3M KCl; 100 mL
9995951

₹6,500.00INR / Each
+
Jenway 4510 Conductivity/TDS Meter and Probe Stand; 120 VAC
3544002

₹106,000.00INR / Each
=
Total Price 722000.00 ADD 9 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Combination electrodes
  • Application specific electrodes
  • Waterproof electrodes
  • Temperature and reference electrodes

More About this Item

Choose the right electrode for your application.

Specifications
  • ApplicationsTest tubes, microfuge tubes, and Tris buffers/solutions
  • Electrode TypeCombination
  • Electrode Body MaterialGlass
  • Reference JunctionFrit ceramic
  • Reference ElectrolyteDouble junction Ag/AgCl
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)50
  • ConnectorBNC
  • Diameter (mm)6
  • Length (mm)90
  • Cable Length (m)1
  • DescriptionSemi-Micro, 90 mm Reach, Glass Bodied Tris Buffer pH Electrode for Biological Buffers, Blood, and Protein Samples
Reviews
Q&A