Jenway Benchtop Conductivity Meters
Quickly get accurate conductivity readings
  • Always measure at the best resolution with automatic standard recognition
  • Easily calibrate with either standard solution or known cell constant
Benchtop conductivity meters offers ±0.5% ±2 digit accuracy with 1, 2 or 3 point conductivity calibration across a wide measurement range. Setup options include cell constant, temperature coefficient, and reference temperature.
Advanced meters help ensure compliance with Good Laboratory Practice (GLP) guidelines with calibration reminder interval, alarm output, and security code setup features. Select from standard and advanced meters are available as meter only or as a kit that includes a conductivity probe with ATC.
106,000.00 - ₹223,000.00INR / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Description
Availability Pricing
COMPARE
230 Conductivity Meter Only; 230 VAC, UK Plug
₹106,000.00INR / Each
COMPARE
230 Conductivity/TDS Meter Kit; 230 VAC/UK
₹123,000.00INR / Each
COMPARE
120 Conductivity/TDS Meter and Probe Stand; 120 VAC
₹106,000.00INR / Each
COMPARE
120 Conductivity/TDS Meter Kit; 120 VAC
₹123,000.00INR / Each
COMPARE
230 Conductivity/TDS Meter Only; 230 VAC, EU Plug
₹106,000.00INR / Each
COMPARE
230 Conductivity/TDS Meter Kit; 230 VAC/EU
₹123,000.00INR / Each
COMPARE
230 Conductivity/TDS/Resistivity/Salinity Meter with GLP and Probe Stand; 230 VAC/UK
₹223,000.00INR / Each
COMPARE
230 Conductivity/TDS/Resistivity/Salinity Meter Kit with GLP; 230 VAC/UK
₹172,000.00INR / Each
COMPARE
120 Conductivity/TDS/Resistivity/Salinity Meter Only with GLP; 120 VAC
₹145,000.00INR / Each
COMPARE
120 Conductivity/TDS/Resistivity/Salinity Meter Kit with GLP; 120 VAC
₹172,000.00INR / Each
COMPARE
230 Conductivity/TDS/Resistivity/Salinity Meter with GLP and Probe Stand; 230 VAC/EU
₹145,000.00INR / Each
COMPARE
230 Conductivity/TDS/Resistivity/Salinity Meter Kit with GLP; 230 VAC/EU
₹173,000.00INR / Each
Showing 1- 12 of 12