Jenway 924 051 Epoxy-Bodied, Flat Membrane pH Electrode for Food Extracts and Agar Plates

Representative image only.

Jenway 924 051 Epoxy-Bodied, Flat Membrane pH Electrode for Food Extracts and Agar Plates

Jenway | Item: 9996043

A variety of pH electrodes for your Jenway meters

₹28,700.00INR / Each

₹28,700.00INR / Each

Buy It Together

Current Item

Jenway 924 051 Epoxy-Bodied, Flat Membrane pH Electrode for Food Extracts and Agar Plates
Jenway 924 051 Epoxy-Bodied, Flat Membrane pH Electrode for Food Extracts and Agar Plates
9996043

₹28,700.00INR / Each
+ + +
Jenway 3510 Standard Digital pH Meter Only, ATC; 230 VAC/UK
3545901

₹100,000.00INR / Each
+ +
Jenway 025 157 Redox Standard, 200 mV; 500 mL
7900029

₹16,200.00INR / Each
+
Jenway 3510 pH Meter with FREE Electrode, 120 VAC
3545913

₹244,000.00INR / Each
+
Jenway 4510 Conductivity/TDS Meter and Probe Stand; 120 VAC
3544002

₹106,000.00INR / Each
=
Total Price 711500.00 ADD 8 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Combination electrodes
  • Application specific electrodes
  • Waterproof electrodes
  • Temperature and reference electrodes

More About this Item

Choose the right electrode for your application.

Specifications
  • ApplicationsFood extracts and viscous samples
  • Electrode TypeCombination
  • Electrode Body MaterialEpoxy
  • Reference JunctionPorous PTFE
  • Reference ElectrolyteAgCl, partial gel filled single junction
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • ConnectorBNC
  • Diameter (mm)12
  • Length (mm)120
  • Cable Length (m)1
  • DescriptionEpoxy-Bodied, Flat Membrane pH Electrode for Food Extracts and Agar Plates
Reviews
Q&A