Mettler Toledo Crucibles for DSC and TGA Measurements
Provides outstanding thermal analysis for a variety samples
  • Large volume crucibles offer increase the sensitivity
  • Low mass crucibles push the resolution to the limit
  • No reactions between sample and crucible caused by impure materials
  • Designed to guarantee high thermal conductivity and dimensional stability
These crucibles are designed for use in Differential Scanning Calorimetry (DSC) and Thermogravimetric Analysis (TGA) to ensure accurate thermal analysis by preventing direct contact between the sample and the sensor, avoiding sensor contamination.
Available in various sizes and materials, they support a wide range of applications, including high-pressure tests, polymer melting, crystallization, and glass transition studies, helping you achieve optimal results in thermal measurements.
94,200.00 - ₹820,000.00INR / Pkg of 20
28 variations of this product are available.
See All Product Options Below
28 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Overall Height (mm) Capacity (mL) Material
Availability Pricing
Mettler Toledo Crucible, Aluminum Oxide, 70 uL; 20/PK 2575738 Mfr # 24123
COMPARE
4.5 0.07 Aluminum Oxide
₹162,000.00INR / Pkg of 20
Mettler Toledo Crucible, Aluminum Oxide, 150 uL; 20/PK 2575739 Mfr # 24124
COMPARE
4.5 0.15 Aluminum Oxide
₹301,000.00INR / Pkg of 20
Mettler Toledo Crucible, Platinum, 150 uL; 4/PK 2575740 Mfr # 24126
COMPARE
4.2 0.15 Platinum
₹744,000.00INR / Pkg of 4
Mettler Toledo High-Pressure Crucible, Gold-Plated Stainless Steel, 40 uL; 25/PK 2575741 Mfr # 26731
COMPARE
5.9 0.04 Gold-Plated Stainless Steel
₹700,000.00INR / Pkg of 25
Mettler Toledo High-Pressure Crucible with Pin, Gold-Plated Stainless Steel, 40 uL; 25/PK 2575742 Mfr # 26732
COMPARE
5.9 0.04 Gold-Plated Stainless Steel
₹820,000.00INR / Pkg of 25
Mettler Toledo Crucible, Aluminum, 40 uL; 100/PK 2575743 Mfr # 26763
COMPARE
1.6 0.04 Aluminum
₹118,000.00INR / Pkg of 100
Mettler Toledo Medium Pressure Crucible with Pin, Staineless Steel, 120 uL; 25/PK 2575744 Mfr # 26929
COMPARE
5.5 0.12 Stainless Steel
₹287,000.00INR / Pkg of 25
Mettler Toledo Crucible, Gold, 40 uL; 6/PK 2575745 Mfr # 27220
COMPARE
1.65 0.04 Gold
₹631,000.00INR / Pkg of 6
Mettler Toledo Crucible wth Pin, Aluminum, 40 uL; 100/PK 2575746 Mfr # 27331
COMPARE
1.6 0.04 Aluminum
₹159,000.00INR / Pkg of 100
Mettler Toledo Crucible with Pin, Aluminum, 160 uL;  40/PK 2575747 Mfr # 27811
COMPARE
6.4 0.16 Aluminum
₹179,000.00INR / Pkg of 40
Mettler Toledo Medium Pressure Crucible, Staineless Steel, 120 uL; 25/PK 2575749 Mfr # 29990
COMPARE
5.5 0.12 Stainless Steel
₹243,000.00INR / Pkg of 25
Mettler Toledo High-Pressure Crucible, Gold-Plated Stainless Steel, 25 uL; 25/PK 2575750 Mfr # 30077139
COMPARE
4.7 0.025 Gold-Plated Stainless Steel
₹412,000.00INR / Pkg of 25
Mettler Toledo Crucible, Aluminum Oxide, 600 uL; 4/PK 2575751 Mfr # 30077260
COMPARE
9.2 0.6 Aluminum Oxide
₹162,000.00INR / Pkg of 4
Mettler Toledo Crucible, Aluminum, 25 uL; 100/PK 2575752 Mfr # 30085850
COMPARE
1.6 0.025 Aluminum
₹94,200.00INR / Pkg of 100
Mettler Toledo Crucible, Aluminum Oxide, 300 uL; 10/PK 2575754 Mfr # 30267108
COMPARE
9 0.3 Aluminum Oxide
₹188,000.00INR / Pkg of 10
Mettler Toledo Crucible, Platinum, 70 uL; 4/PK 2575755 Mfr # 51119654
COMPARE
4.2 0.07 Platinum
₹537,000.00INR / Pkg of 4
Mettler Toledo Crucible, Light Aluminum, 20 uL; 100/PK 2575756 Mfr # 51119810
COMPARE
1.6 0.2 Aluminum
₹181,000.00INR / Pkg of 100
Mettler Toledo Pan, Aluminum, 40 uL; 400/PK 2575757 Mfr # 51119870
COMPARE
1.6 0.04 Aluminum
₹362,000.00INR / Pkg of 400
Mettler Toledo Pan, Aluminum, 100 uL; 400/PK 2575758 Mfr # 51119872
COMPARE
4.2 0.1 Aluminum
₹626,000.00INR / Pkg of 400
Mettler Toledo Crucible, Aluminum Oxide, 900 uL; 4/PK 2575759 Mfr # 51119960
COMPARE
10 0.9 Aluminum Oxide
₹181,000.00INR / Pkg of 4
Showing 1 - 20 of 28