Mettler Toledo Medium Pressure Crucible, Staineless Steel, 120 uL; 25/PK
Representative image only.
Mettler Toledo Medium Pressure Crucible, Staineless Steel, 120 uL; 25/PK
Mettler Toledo | Mfr: 29990| Item: 2575749
Provides outstanding thermal analysis for a variety samples
₹243,000.00INR / Pkg of 25
₹243,000.00INR / Pkg of 25
Product Description
Includes
Lid.
Key Features
  • Large volume crucibles offer increase the sensitivity
  • Low mass crucibles push the resolution to the limit
  • No reactions between sample and crucible caused by impure materials
  • Designed to guarantee high thermal conductivity and dimensional stability
  • Recommended for DSC measurements
  • Can withstand maximum pressure is 2 Mpa
  • Can be used without O-ring
More About this Item
These crucibles are designed for use in Differential Scanning Calorimetry (DSC) and Thermogravimetric Analysis (TGA) to ensure accurate thermal analysis by preventing direct contact between the sample and the sensor, avoiding sensor contamination.
Available in various sizes and materials, they support a wide range of applications, including high-pressure tests, polymer melting, crystallization, and glass transition studies, helping you achieve optimal results in thermal measurements.
This medium-pressure crucible is ideal for samples containing liquids such as water, for temperatures above boiling point. Please note, when sealed with a lid and special seal, it is a single use crucible. It cannot be regenerated.
Specifications
  • MaterialStainless Steel
  • Capacity (mL)0.12
  • Cap TypeLid
  • Cap MaterialStainless steel
  • Overall Height (mm)5.5
  • Bottom Outside Diameter (mm)6
  • DescriptionMedium Pressure Crucible, Staineless Steel, 120 uL; 25/PK