Mettler Toledo Crucible with Pin, Aluminum, 160 uL; 40/PK
Representative image only.
Mettler Toledo Crucible with Pin, Aluminum, 160 uL; 40/PK
Mettler Toledo | Mfr: 27811| Item: 2575747
Provides outstanding thermal analysis for a variety samples
₹179,000.00INR / Pkg of 40
₹179,000.00INR / Pkg of 40
Product Description
Includes
Lid and pin.
Key Features
  • Large volume crucibles offer increase the sensitivity
  • Low mass crucibles push the resolution to the limit
  • No reactions between sample and crucible caused by impure materials
  • Designed to guarantee high thermal conductivity and dimensional stability
  • Recommended for DSC measurements
  • Flat base ensures excellent thermal contact
  • Can be used without furnace expander if the lid is reversed
More About this Item
These crucibles are designed for use in Differential Scanning Calorimetry (DSC) and Thermogravimetric Analysis (TGA) to ensure accurate thermal analysis by preventing direct contact between the sample and the sensor, avoiding sensor contamination.
Available in various sizes and materials, they support a wide range of applications, including high-pressure tests, polymer melting, crystallization, and glass transition studies, helping you achieve optimal results in thermal measurements.
Specifications
  • MaterialAluminum
  • Capacity (mL)0.16
  • Cap TypeLid
  • Cap MaterialAluminum
  • Overall Height (mm)6.4
  • Bottom Outside Diameter (mm)6
  • DescriptionCrucible with Pin, Aluminum, 160 uL; 40/PK