Mettler Toledo High-Pressure Crucible with Pin, Gold-Plated Stainless Steel, 40 uL; 25/PK
Representative image only.
Mettler Toledo High-Pressure Crucible with Pin, Gold-Plated Stainless Steel, 40 uL; 25/PK
Mettler Toledo | Mfr: 26732| Item: 2575742
Provides outstanding thermal analysis for a variety samples
₹820,000.00INR / Pkg of 25
₹820,000.00INR / Pkg of 25
Product Description
Includes
Lid and pin.
Key Features
  • Large volume crucibles offer increase the sensitivity
  • Low mass crucibles push the resolution to the limit
  • No reactions between sample and crucible caused by impure materials
  • Designed to guarantee high thermal conductivity and dimensional stability
  • Can withstand an internal pressure of 15 Mpa
  • Ideal for use with highly reactive materials
  • Good for one time use
More About this Item
These crucibles are designed for use in Differential Scanning Calorimetry (DSC) and Thermogravimetric Analysis (TGA) to ensure accurate thermal analysis by preventing direct contact between the sample and the sensor, avoiding sensor contamination.
Available in various sizes and materials, they support a wide range of applications, including high-pressure tests, polymer melting, crystallization, and glass transition studies, helping you achieve optimal results in thermal measurements.
Specifications
  • MaterialGold-Plated Stainless Steel
  • Capacity (mL)0.04
  • Cap TypeLid
  • Cap MaterialGold-plated stainles steel
  • Overall Height (mm)5.9
  • Bottom Outside Diameter (mm)7
  • DescriptionHigh-Pressure Crucible with Pin, Gold-Plated Stainless Steel, 40 uL; 25/PK