Cole-Parmer Vacuum Filtration Holder; membrane size, 25mm

Representative image only.

Cole-Parmer Vacuum Filtration Holder; membrane size, 25mm

Cole-Parmer | Item: 0292000

Optimize your vacuum filtration process

₹61,400.00INR / Each

₹61,400.00INR / Each

Buy It Together

Current Item

Cole-Parmer Vacuum Filtration Holder; membrane size, 25mm
Cole-Parmer Vacuum Filtration Holder; membrane size, 25mm
0292000

₹61,400.00INR / Each
+ + + + + + + +
Cole-Parmer Filtering Flask; 1000 mL
3450912

₹19,700.00INR / Each
=
Total Price 1481200.00 ADD 9 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Model 02923-20 meets EPA specifications for Hazardous Waste Toxicity Test
  • Autoclavable
  • Supplied with an aluminum clamp and a silicone rubber stopper
  • 1 year warranty

More About this Item

No. 5 stoppers fit standard 125-mL flasks; no. 8 stoppers fit 1-L flasks. Order no. 8B stopper 02924-02 separately at right to fit filter holders 02920-00 and 02921-00 to manifolds above. Filter holder 02923-10 is ideal for sterility testing of antibiotics and pharmaceuticals by bacteria retentive method described in USP XXI. This model includes funnel, filter base, aluminum clamp, and stopper. Model 02923-20 with sintered-glass membrane support meets EPA specifications for use in the Hazardous Waste Toxicity Test. Holder 02923-00, with stainless steel (SS) membrane support, provides ultraclean filtrate—screen does not shed particles.

To fit holder to manifold, order no. 8B stopper 02924-02 separately.

1 Year Warranty
Specifications
  • Diameter (mm)25
  • Diameter (cm)2.5
  • Funnel Size (mL)100
  • Membrane SupportSintered Glass
  • Stopper NumberNo. 5
  • Filtration Area (cm²)1.1
  • DescriptionVacuum Filtration Holder; membrane size, 25mm
  • Warranty1 year
Reviews
Q&A