Cole-Parmer® Filtration Assembly
Ideal for filtering small to medium volumes of liquid
  • Sintered glass support provides an even, flat support structure for membrane filters
  • Minimize contact between sample and filtration system
  • Complete filtration assembly — just add a membrane
These Cole-Parmer glass filtration assemblies will accept a 47-mm membrane and will fit perfectly into a filter flask with no. 8 stopper neck size. Glass filter holder assembly consists of a 300-mL borosilicate glass funnel and base, no. 8 silicone rubber stopper with hole, and anodized aluminum clamp.
These filtration assemblies are designed for HPLC, particulate, and microbiological contamination filtrations of particulate analysis.
23,300.00 - ₹31,700.00INR / Set of 1
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Filter Diameter (mm) Funnel Size (mL) Description
Availability Pricing
COMPARE
47 300 Glass Filtration Assembly, 47 mm Diameter, 300 mL; Stopper with 9/16\" Diameter Hole
₹23,300.00INR / Set of 1
COMPARE
47 300 Glass Filtration Assembly with 1000 mL Flask, 47 mm Diameter, 300 mL; Stopper with 9/16\" Diameter Hole
₹31,700.00INR / Set of 1