Cole-Parmer® Glass Filtering Flasks
Ideal for vacuum filtration
  • Constructed of durable glass for enhanced mechanical strength
Cole-Parmer filtration flasks feature tubulation which allows for easy connection to 3/8” ID vacuum tubing. The neck finish affords an excellent fit for number 8B rubber stoppers or Cole-Parmer filtration assemblies. Order Cole-Parmer filtration assemblies separately.
9,800.00 - ₹14,700.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL)
Availability Pricing
COMPARE
1000
₹9,800.00INR / Each
COMPARE
2000
₹14,700.00INR / Each