Advantec All-Glass Filtration Assemblies
Ideal for high-purity and aggressive chemical applications
All-glass assemblies are ideal for aggressive solvents and applications where you need to minimize the materials that contact sample or filtrate. Standard 25- and 47-mm holders mount on filtration flask using a ground glass joint. Outlet of support base drip tube is positioned below the side arm connection to prevent sample aspiration into vacuum line.
225,000.00 - ₹373,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
RESOURCES
+ SAVE TO MY DOCS (PDFs)
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Filter Diameter (mm) Receiver Capacity (mL) Support
Availability Pricing
Advantec 311220 All-Glass Filtration Assembly for 25 mm Membranes; 15 mL 0664519 Mfr # 311220
COMPARE
25 250 Glass Frit
Call For Price
Advantec 311320 Glass Filtration Assembly for 25mm Membranes w/SS Support; 15 mL 0664539 Mfr # 311320
COMPARE
25 250 Stainless Steel
₹225,000.00INR / Each
Advantec 311620 Glass Filtration Assembly for 47mm Membranes w/PTFE Support; 300 mL 0664541 Mfr # 311620
COMPARE
47 1000 Stainless Steel
₹373,000.00INR / Each
Advantec 311420 All-Glass Filtration Assembly for 47 mm Membranes; 300 mL 0664549 Mfr # 311420
COMPARE
47 1000 Stainless Steel
₹319,000.00INR / Each
Advantec 311520 Glass Filtration Assembly for 47mm Membranes w/SS Support; 30 mL 0664565 Mfr # 311520
COMPARE
47 1000 Stainless Steel
₹290,000.00INR / Each