Advantec Standard Glass Microanalysis Filter Holders
Ideal for small to medium volumes of liquid
  • Standard sizes for microbiology and particulate analysis
  • Select glass support for low pH solutions or for slower flow rates; use stainless for proteinaceous or viscous solutions; use PTFE support for acidic solutions at higher flow rates
Microanalysis filter holders feature borosilicate glass funnel and base, silicone rubber stopper, and anodized aluminum clamp. The 90-mm holders are ideal for large-volume viscous samples or those with a heavy particle or bioburden that would clog a 47-mm filter.
104,000.00 - ₹448,000.00INR / Each
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Body Material Filter Diameter (mm) Support
Availability Pricing
Advantec 311000 All-Glass Microanalysis Filter Holder, 13 mm; 100 mL 0664481 Mfr # 311000
COMPARE
Glass 13 Sintered Glass
₹126,000.00INR / Each
Advantec 311050 All-Glass Microanalysis Filter Holder, 13 mm; 500 mL 0664482 Mfr # 311050
COMPARE
Glass 13 Sintered Glass
₹161,000.00INR / Each
Advantec 311110 All-Glass Microanalysis Filter Holder, 13 mm; 1000 mL 0664483 Mfr # 311110
COMPARE
Glass 13 Sintered Glass
₹173,000.00INR / Each
Advantec 311100 All-Glass Microanalysis Filter Holder, 13 mm; 13 or 100 mL 0664484 Mfr # 311100
COMPARE
Glass 13 Sintered Glass
₹167,000.00INR / Each
Advantec 311400 All-Glass Microanalysis Filter Holder, 47 mm; 300 mL 0664485 Mfr # 311400
COMPARE
Glass 47 Sintered Glass
₹104,000.00INR / Each
Advantec 311500 Glass Microanalysis Filter Holder, 47 mm, SS Support; 300 ml 06644-86 0664486 Mfr # 311500
COMPARE
Stainless Steel 47 Stainless Steel
₹191,000.00INR / Each
Advantec 311540 Glass Microanalysis Filter Holder, 47 mm, PTFE Support; 300 mL 0664487 Mfr # 311540
COMPARE
PTFE 47 PTFE
₹221,000.00INR / Each
Advantec 311480 All-Glass Microanalysis Filter Holder, 47 mm; 100 mL 0664488 Mfr # 311480
COMPARE
PTFE 47 Sintered Glass
₹172,000.00INR / Each
Advantec 311680 Glass Microanalysis Filter Holder, 47 mm, PTFE Support; 100 mL 0664490 Mfr # 311680
COMPARE
PTFE 47 PTFE
₹225,000.00INR / Each
Advantec 311490 All-Glass Microanalysis Filter Holder for 47 mm Membranes; 500 mL 0664491 Mfr # 311490
COMPARE
Glass 47 Sintered Glass
₹197,000.00INR / Each
Advantec 311590 Glass Microanalysis Filter Holder, 47 mm, SS Support; 500 mL 0664492 Mfr # 311590
COMPARE
Stainless Steel 47 Stainless Steel
₹225,000.00INR / Each
Advantec 311690 Glass Microanalysis Filter Holder, 47 mm, PTFE Support; 500 mL 0664493 Mfr # 311690
COMPARE
PTFE 47 PTFE
₹258,000.00INR / Each
Advantec 314110 All-Glass Microanalysis Filter Holder, 47 mm; 1000 mL 0664494 Mfr # 314110
COMPARE
Glass 47 Sintered Glass
₹240,000.00INR / Each
Advantec 316110 Glass Microanalysis Filter Holder, 47 mm, PTFE Support; 1000 mL 0664496 Mfr # 316110
COMPARE
PTFE 47 PTFE
₹252,000.00INR / Each
Advantec 352100 All-Glass Microanalysis Filter Holder, 90 mm; 1000 mL 0664497 Mfr # 352100
COMPARE
Glass 90 Sintered Glass
₹448,000.00INR / Each
Advantec 352200 Glass Microanalysis Filter Holder, 90 mm, SS Support; 1000 mL 0664498 Mfr # 352200
COMPARE
Stainless Steel 90 Stainless Steel
₹311,000.00INR / Each
Advantec 311200 All-Glass Microanalysis Filter Holder, 25 mm; 15 mL 0664513 Mfr # 311200
COMPARE
Glass 25 Sintered Glass
₹129,000.00INR / Each
Advantec 311280 All-Glass Microanalysis Filter Holder, 25 mm; 150 mL 0664523 Mfr # 311280
COMPARE
Glass 25 Sintered Glass
₹147,000.00INR / Each
Advantec 311290 All-Glass Microanalysis Filter Holder, 25 mm; 300 mL 0664525 Mfr # 311290
COMPARE
Glass 25 Sintered Glass
₹162,000.00INR / Each
Advantec 311380 Glass Microanalysis Filter Holder, 25 mm, SS Support; 150 mL 0664596 Mfr # 311380
COMPARE
Glass 25 Stainless Steel
₹167,000.00INR / Each
Showing 1- 20 of 20