Chemetrics K-2505 Colorimetric Chlorine D DPD Test Kit
Representative image only.
Chemetrics K-2505 Colorimetric Chlorine D DPD Test Kit
Chemetrics | Mfr: K-2505| Item: 0554042
CHEMets test kits are portable and self-contained
₹45,500.00INR / Each
₹45,500.00INR / Each
Buy It Together
Current Item
Chemetrics K-2505 Colorimetric Chlorine D DPD Test Kit
Chemetrics K-2505 Colorimetric Chlorine D DPD Test Kit
0554042
₹45,500.00INR / Each
+ + +
Chemetrics R-2505 Refill Kit for 05540-42, 30 Tests
0554043
₹17,000.00INR / Each
+ =
Total Price 111000.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Vacuum sealed reagents and handy comparator chart for quick, visual results
  • 2 year warranty
More About this Item
CHEMETS colorimetric test kits let you visually compare the color of your sample to standards using a comparator. Just break tip of ampoule in cup (included) filled with sample. Vacuum pulls sample into ampoule automatically; gently swirl to mix. Handy color comparator (included) provides accurate visual analysis. Vacuum packing keeps reagents active indefinitely.
The chlorine DDPD test kit measures free and total chlorine.
Two year warranty
Specifications
  • TestChlorine, free and total
  • Measurement Points0 to 1 ppm; 1 to 5 ppm
  • Reagent PlatformDDPD Method
  • Tests Per Kit30
  • DescriptionColorimetric Chlorine D DPD Test Kit