CHEMets Colorimetric Dissolved Oxygen Test Kits
Vacuum sealed reagents and handy comparator chart for quick, visual results
Just break tip of ampoule in cup (included) filled with sample. Vacuum pulls sample into ampoule automatically; gently swirl to mix. Handy color comparator (included) provides accurate visual analysis. Vacuum packing keeps reagents active indefinitely.
26,800.00 - ₹29,300.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Measurement Points Test
Availability Pricing
Chemetrics K-7540 Colorimetric Dissolved Oxygen Test Kit , 0 to 40 ppb 0554020 Mfr # K-7540
COMPARE
0 to 40 ppb Dissolved oxygen
₹29,300.00INR / Each
Chemetrics K-7599 Colorimetric Dissolved Oxygen Test Kit , 0 to 100 ppb 0554025 Mfr # K-7599
COMPARE
0 to 100 ppb Dissolved oxygen
₹29,300.00INR / Each
Chemetrics K-7501 Colorimetric Dissolved Oxygen Test Kit , 0 to 1 ppm 0554030 Mfr # K-7501
COMPARE
0 to 1 ppm Dissolved oxygen
₹26,800.00INR / Each
Chemetrics K-7512 Colorimetric Dissolved Oxygen Test Kit , 1 to 12 ppm 0554040 Mfr # K-7512
COMPARE
1 to 12 ppm Dissolved oxygen
₹26,800.00INR / Each