Chemetrics K-7512 Colorimetric Dissolved Oxygen Test Kit , 1 to 12 ppm
Representative image only.
Chemetrics K-7512 Colorimetric Dissolved Oxygen Test Kit , 1 to 12 ppm
Chemetrics | Mfr: K-7512| Item: 0554040
Vacuum sealed reagents and handy comparator chart for quick, visual results
₹26,800.00INR / Each
₹26,800.00INR / Each
Buy It Together
Current Item
Chemetrics K-7512 Colorimetric Dissolved Oxygen Test Kit , 1 to 12 ppm
Chemetrics K-7512 Colorimetric Dissolved Oxygen Test Kit , 1 to 12 ppm
0554040
₹26,800.00INR / Each
+
Chemetrics K-3510 Colorimetric Copper Test Kit
0029050
Call For Price
+ + =
Total Price 57700.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Includes
polypropylene case with instructions; all necessary apparatus; and reagent vials for 30 tests.
More About this Item
Just break tip of ampoule in cup (included) filled with sample. Vacuum pulls sample into ampoule automatically; gently swirl to mix. Handy color comparator (included) provides accurate visual analysis. Vacuum packing keeps reagents active indefinitely.
Specifications
  • TestDissolved oxygen
  • Measurement Points1 to 12 ppm
  • Tests Per Kit30
  • DescriptionColorimetric Dissolved Oxygen Test Kit , 1 to 12 ppm