Titrets Titrimetric Test Kits
Includes the Titrettor™ device for precise sample control and instant titrimetric analysis
  • No mess with premeasured reagent ampoules which are inverted after testing to show your results
Perform most tests in three minutes or less—ideal for field or lab use. Repeatable procedure is even straightforward enough for nontechnical personnel. Test kits include all necessary apparatus.
Test kits let you control the amount of sample drawn into the vial. Once a color change occurs, you can check the liquid level in the vial to determine sample concentration in ppm.
Kits can be stored indefinitely (except the calcium hardness test kit, which has a shelf life of 7 months and the chloride test kits, which have a shelf life of 4 months).
What's included: instructions, all necessary apparatus, and reagent vials for 30 tests. Refill kits contain 30 vials each.
13,000.00 - ₹14,300.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Measurement Points Test Tests Per Kit
Availability Pricing
Chemetrics K-9810 Titrimetric Total Alkalinty, Lr, Test Kits WW-05540-66 Mfr # K-9810
COMPARE
10 to 100 ppm Alkalinity, total 30
₹14,300.00INR / Each
Chemetrics K-9815 Titrimetric Total Alkalinty, Hr, Test Kits WW-05540-68 Mfr # K-9815
COMPARE
50 to 500 ppm Alkalinity, total 30
₹13,400.00INR / Each
Chemetrics K-1705 Titrimetric Calcium Hardness Test Kits WW-05540-70 Mfr # K-1705
COMPARE
50 to 500 ppm Hardness, calcium 30
₹13,400.00INR / Each
Chemetrics K-1910 Titrimetric Carbon Dioxide Test Kits WW-05540-72 Mfr # K-1910
COMPARE
10 to 100 ppm Carbon dioxide 30
Call For Price
Chemetrics K-4502 Titrimetric Hardness, 2 to 20 ppm, Test Kits WW-05540-78 Mfr # K-4502
COMPARE
2 to 20 ppm Hardness, total 30
₹13,400.00INR / Each
Chemetrics K-4520 Titrimetric Hardness, 20 to 200 ppm, Test Kits WW-05540-80 Mfr # K-4520
COMPARE
20 to 200 ppm Hardness, total 30
₹13,000.00INR / Each
Chemetrics K-4585 Titrimetric Hardness, 100 to 1000 ppm, Test Kits WW-05540-82 Mfr # K-4585
COMPARE
100 to 1000 ppm Hardness, total 30
₹13,000.00INR / Each
Chemetrics K-9602 Titrimetric Sulfite, 2 to 20 ppm, Test Kits WW-05540-84 Mfr # K-9602
COMPARE
2 to 20 ppm Sulfite 30
Call For Price
Chemetrics K-9605 Titrimetric Sulfite, 5 to 50 ppm, Test Kits WW-05540-86 Mfr # K-9605
COMPARE
5 to 50 ppm Sulfite 30
Call For Price
Chemetrics K-9610 Titrimetric Sulfite, 10 to 100 ppm, Test Kits WW-05540-88 Mfr # K-9610
COMPARE
10 to 100 ppm Sulfite 30
Call For Price
Showing 1- 10 of 10