Chemetrics K-4585 Titrimetric Hardness, 100 to 1000 ppm, Test Kits
Representative image only.
Chemetrics K-4585 Titrimetric Hardness, 100 to 1000 ppm, Test Kits
Chemetrics | Mfr: K-4585| Item: 0554082
Includes the Titrettor™ device for precise sample control and instant titrimetric analysis
₹16,000.00INR / Each
₹16,000.00INR / Each
Buy It Together
Current Item
Chemetrics K-4585 Titrimetric Hardness, 100 to 1000 ppm, Test Kits
Chemetrics K-4585 Titrimetric Hardness, 100 to 1000 ppm, Test Kits
0554082
₹16,000.00INR / Each
+ + =
Total Price 56100.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • No mess with premeasured reagent ampoules which are inverted after testing to show your results
  • 1 year warranty
More About this Item
Perform most tests in three minutes or less—ideal for field or lab use. Repeatable procedure is even straightforward enough for nontechnical personnel. Test kits include all necessary apparatus.
Test kits let you control the amount of sample drawn into the vial. Once a color change occurs, you can check the liquid level in the vial to determine sample concentration in ppm.
Kits can be stored indefinitely (except the calcium hardness test kit, which has a shelf life of 7 months and the chloride test kits, which have a shelf life of 4 months).
What's included: instructions, all necessary apparatus, and reagent vials for 30 tests. Refill kits contain 30 vials each.
1 Year Warranty
Specifications
  • TestHardness, total
  • Measurement Points100 to 1000 ppm
  • Tests Per Kit30
  • DescriptionTitrimetric Hardness, 100 to 1000 ppm, Test Kits