Chemetrics R-7512 Refill kit for 05540-40, 30/box (Dissolved oxygen test refill)
Representative image only.
Chemetrics R-7512 Refill kit for 05540-40, 30/box (Dissolved oxygen test refill)
Chemetrics | Mfr: R-7512| Item: 0554041
Take it wherever you go
₹13,300.00INR / Box of 30
₹13,300.00INR / Box of 30
Buy It Together
Current Item
Chemetrics R-7512 Refill kit for 05540-40, 30/box (Dissolved oxygen test refill)
Chemetrics R-7512 Refill kit for 05540-40, 30/box (Dissolved oxygen test refill)
0554041
₹13,300.00INR / Box of 30
+ +
Chemetrics K-3510 Colorimetric Copper Test Kit
0029050
Call For Price
+ =
Total Price 57700.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Vacuum sealed packaging keeps reagents active
More About this Item
CHEMets colorimetric test kits let you visually compare the color of your sample to standards using a comparator, comparator sold separately. Just break tip of ampoule in cup filled with sample. Vacuum pulls sample into ampoule automatically; gently swirl to mix. Handy color comparator provides accurate visual analysis.
Specifications
  • Compatible With05540-40
  • DescriptionRefill kit for 05540-40, 30/box (Dissolved oxygen test refill)