CHEMets Colorimetric Visual Test Kits
CHEMets test kits are portable and self-contained
  • Vacuum sealed reagents and handy comparator chart for quick, visual results
CHEMETS colorimetric test kits let you visually compare the color of your sample to standards using a comparator. Just break tip of ampoule in cup (included) filled with sample. Vacuum pulls sample into ampoule automatically; gently swirl to mix. Handy color comparator (included) provides accurate visual analysis. Vacuum packing keeps reagents active indefinitely.
29,800.00 - ₹45,500.00INR / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Measurement Points Test Description
Availability Pricing
Chemetrics K-8012 Phenol CHEMets Visual Kit 0029030 Mfr # K-8012
COMPARE
0 to 1 ppm; 0 to 12 ppm Phenol Phenol CHEMets Visual Kit
₹31,600.00INR / Each
Chemetrics K-3510 Colorimetric Copper Test Kit 0029050 Mfr # K-3510
COMPARE
0 to 1 ppm; 1 to 10 ppm Copper, total Colorimetric Copper Test Kit
Call For Price
Chemetrics K8515 Colorimetric Phosphate, Hr, Test Kit 0029052 Mfr # K8515
COMPARE
10 to 150 ppm Phosphate Colorimetric Phosphate, Hr, Test Kit
Call For Price
Chemetrics K-1420 Colorimetric Ammonia/Nitrogen Test Kit 0029100 Mfr # K-1420
COMPARE
0 to 4 ppm; 0 to 80 ppm Ammonia, nitrogen Colorimetric Ammonia/Nitrogen Test Kit
Call For Price
Chemetrics K-6010 Colorimetric Iron Test Kit 0029140 Mfr # K-6010
COMPARE
0 to 1 ppm; 1 to 10 ppm Iron, total soluble Colorimetric Iron Test Kit
Call For Price
Chemetrics K-9400 Colorimetric Detergent Test Kit 0554000 Mfr # K-9400
COMPARE
0 to 3 ppm Detergents Colorimetric Detergent Test Kit
Call For Price
Chemetrics K-2505 Colorimetric Chlorine D DPD Test Kit 0554042 Mfr # K-2505
COMPARE
0 to 1 ppm; 1 to 5 ppm Chlorine, free and total Colorimetric Chlorine D DPD Test Kit
₹45,500.00INR / Each
Chemetrics K-9510 Colorimetric Sufide Test Kit 0554052 Mfr # K-9510
COMPARE
0 to 1 ppm; 1 to 10 ppm Sulfide, total soluble Colorimetric Sufide Test Kit
Call For Price
Chemetrics K-2504 Test Kit Chlorine 0 to 1, 1 to 5 ppm Range s 0554090 Mfr # K-2504
COMPARE
0 to 1 ppm; 1 to 5 ppm Chlorine, free and total Test Kit Chlorine 0 to 1, 1 to 5 ppm Range s
₹29,800.00INR / Each
Chemetrics K-8510 Colorimetric Phosphate Test Kit 0554092 Mfr # K-8510
COMPARE
0 to 1 ppm; 1 to 10 ppm Phosphate Colorimetric Phosphate Test Kit
Call For Price
Chemetrics K-5510 Colorimetric Hydrogen Peroxide Test Kit 0554102 Mfr # K-5510
COMPARE
0 to 1 ppm; 1 to 10 ppm Hydrogen peroxide Colorimetric Hydrogen Peroxide Test Kit
₹38,400.00INR / Each
Chemetrics K-9010 Colorimetric Silica Test Kit 0554104 Mfr # K-9010
COMPARE
0 to 1 ppm; 1 to 10 ppm Silica Colorimetric Silica Test Kit
₹32,000.00INR / Each
Showing 1- 12 of 12