Chemetrics K-5510 Colorimetric Hydrogen Peroxide Test Kit
Representative image only.
Chemetrics K-5510 Colorimetric Hydrogen Peroxide Test Kit
Chemetrics | Mfr: K-5510| Item: 0554102
CHEMets test kits are portable and self-contained
₹38,400.00INR / Each
₹38,400.00INR / Each
Buy It Together
Current Item
Chemetrics K-5510 Colorimetric Hydrogen Peroxide Test Kit
Chemetrics K-5510 Colorimetric Hydrogen Peroxide Test Kit
0554102
₹38,400.00INR / Each
+ + +
Chemetrics R-5510 Refill Kit for 05541-02, 30 Tests
0554103
₹15,600.00INR / Each
+ =
Total Price 102500.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Vacuum sealed reagents and handy comparator chart for quick, visual results
  • 2 year warranty
More About this Item
CHEMETS colorimetric test kits let you visually compare the color of your sample to standards using a comparator. Just break tip of ampoule in cup (included) filled with sample. Vacuum pulls sample into ampoule automatically; gently swirl to mix. Handy color comparator (included) provides accurate visual analysis. Vacuum packing keeps reagents active indefinitely.
Two year warranty
Specifications
  • TestHydrogen peroxide
  • Measurement Points0 to 1 ppm; 1 to 10 ppm
  • Tests Per Kit30
  • DescriptionColorimetric Hydrogen Peroxide Test Kit