LaMotte Insta-TEST® Nitrate/Nitrite Test Strips, 0 to 50 ppm Nitrate/0 to 10 Nitrite; 50 Tests Each
Representative image only.
LaMotte Insta-TEST® Nitrate/Nitrite Test Strips, 0 to 50 ppm Nitrate/0 to 10 Nitrite; 50 Tests Each
LaMotte | Mfr: 2996| Item: 9953276
Monitor water quality contaminants with quick spot tests
₹8,400.00INR / Pkg of 25
₹8,400.00INR / Pkg of 25
Buy It Together
Current Item
LaMotte Insta-TEST®  Nitrate/Nitrite Test Strips, 0 to 50 ppm Nitrate/0 to 10 Nitrite; 50 Tests Each
LaMotte Insta-TEST® Nitrate/Nitrite Test Strips, 0 to 50 ppm Nitrate/0 to 10 Nitrite; 50 Tests Each
9953276
₹8,400.00INR / Pkg of 25
+
Chemetrics K-3510 Colorimetric Copper Test Kit
0029050
Call For Price
=
Total Price 8400.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
50 nitrate test strips and 50 nitrite test strips.
Key Features
  • Convenient, on-site water testing with bright easy-to-read colors
  • Pop-top vial with desiccant liner protects strips from moisture
More About this Item
Moisture protection, and fast, reliable results make the Insta-TEST an unbeatable system for water quality testing. Simply dip polystyrene strips into water sample, and compare the resulting color pad change to color chart.
Specifications
  • TestNitrate; Nitrite
  • Measurement Points0, 5, 10, 25, 50; 0, 0.5, 1, 5, 10 ppm
  • Reagent PlatformStrips
  • DescriptionNitrate/Nitrite Test Strips, 0 to 50/0 to 10