LaMotte Insta-TEST Water Test Strips
Monitor water quality contaminants with quick spot tests
  • Convenient, on-site water testing with bright easy-to-read colors
  • Pop-top vial with desiccant liner protects strips from moisture
Moisture protection, and fast, reliable results make the Insta-TEST an unbeatable system for water quality testing. Simply dip polystyrene strips into water sample, and compare the resulting color pad change to color chart.
3,000.00 - ₹8,900.00INR / Pkg of 25
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Measurement Points Test
Availability Pricing
LaMotte Insta-Test Alkalinity Test Strips, 0 to 180 ppm 9953262 Mfr # 2997
COMPARE
0, 40, 80, 120, 180 ppm Alkalinity
₹4,900.00INR / Pkg of 50
LaMotte Insta-Test Ammonia Test Strips, 0 to 6 ppm, 25/Pk 9953264 Mfr # 3023-G
COMPARE
0, 0.5, 1, 3, 6 ppm Ammonia
₹8,900.00INR / Pkg of 25
LaMotte Insta-Test Free Chlorine Test Strips, 0 to 10 ppm 9953266 Mfr # 2964-G
COMPARE
0, 0.5, 1, 3, 5, 10 ppm Chlorine, free
₹3,000.00INR / Pkg of 25
LaMotte Insta-Test Total Chlorine Test Strips, 0 To10 ppm 9953268 Mfr # 2963LR-G
COMPARE
0, 0.25, 0.5, 1, 3, 10 ppm Chlorine, total
Call For Price
LaMotte Insta-Test Free and Total Chlorine Test Strips, 0 to 10 ppm 9953270 Mfr # 3027-G
COMPARE
0, 0.25, 0.5, 1, 3, 5, 10 ppm Chlorine, free and total
₹4,400.00INR / Pkg of 25
LaMotte Insta-Test Total Chlorine Test Strips, 0 to 5 ppm 9953272 Mfr # 2979
COMPARE
0, 0.5, 1, 3, 5 ppm Chlorine, total
₹4,400.00INR / Pkg of 25
LaMotte Insta-Test Hardness Test Strips, 0 to 180 ppm 9953274 Mfr # 2981
COMPARE
0, 30, 60, 120, 180 ppm Hardness
₹5,300.00INR / Pkg of 50
LaMotte Insta-TEST®  Nitrate/Nitrite Test Strips, 0 to 50 ppm Nitrate/0 to 10 Nitrite; 50 Tests Each 9953276 Mfr # 2996
COMPARE
0, 5, 10, 25, 50; 0, 0.5, 1, 5, 10 ppm Nitrate; Nitrite
₹8,400.00INR / Pkg of 25
LaMotte Insta-Test Hydrogen Peroxide Test Strips, 0 to 90 ppm 9953278 Mfr # 2984
COMPARE
0, 15, 30, 50, 90 ppm Hydrogen peroxide
₹3,400.00INR / Pkg of 25
LaMotte Insta-Test Hydrogen Peroxide Test Strips, 0 To50 ppm 9953280 Mfr # 2984LR
COMPARE
0, 1, 3, 10, 50 ppm Hydrogen peroxide
₹5,100.00INR / Pkg of 25
Showing 1- 10 of 10