LaMotte Insta-Test Hydrogen Peroxide Test Strips, 0 to 90 ppm
Representative image only.
LaMotte Insta-Test Hydrogen Peroxide Test Strips, 0 to 90 ppm
LaMotte | Mfr: 2984| Item: 9953278
Monitor water quality contaminants with quick spot tests
₹3,400.00INR / Pkg of 25
₹3,400.00INR / Pkg of 25
Buy It Together
Current Item
LaMotte Insta-Test Hydrogen Peroxide Test Strips, 0 to 90 ppm
LaMotte Insta-Test Hydrogen Peroxide Test Strips, 0 to 90 ppm
9953278
₹3,400.00INR / Pkg of 25
+
Chemetrics K-3510 Colorimetric Copper Test Kit
0029050
Call For Price
=
Total Price 3400.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Convenient, on-site water testing with bright easy-to-read colors
  • Pop-top vial with desiccant liner protects strips from moisture
  • 25 strips per vial.
More About this Item
Moisture protection, and fast, reliable results make the Insta-TEST an unbeatable system for water quality testing. Simply dip polystyrene strips into water sample, and compare the resulting color pad change to color chart.
Specifications
  • TestHydrogen peroxide
  • Measurement Points0, 15, 30, 50, 90 ppm
  • Reagent PlatformStrips
  • DescriptionHydrogen Peroxide Test Strips, 0 to 90 ppm