LaMotte Insta-Test Free Chlorine Test Strips, 0 to 10 ppm
Representative image only.
LaMotte Insta-Test Free Chlorine Test Strips, 0 to 10 ppm
LaMotte | Mfr: 2964-G| Item: 9953266
Monitor water quality contaminants with quick spot tests
₹3,000.00INR / Pkg of 25
₹3,000.00INR / Pkg of 25
Buy It Together
Current Item
LaMotte Insta-Test Free Chlorine Test Strips, 0 to 10 ppm
LaMotte Insta-Test Free Chlorine Test Strips, 0 to 10 ppm
9953266
₹3,000.00INR / Pkg of 25
+
Chemetrics K-3510 Colorimetric Copper Test Kit
0029050
Call For Price
=
Total Price 3000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Convenient, on-site water testing with bright easy-to-read colors
  • Pop-top vial with desiccant liner protects strips from moisture
  • 25 strips per vial.
More About this Item
Moisture protection, and fast, reliable results make the Insta-TEST an unbeatable system for water quality testing. Simply dip polystyrene strips into water sample, and compare the resulting color pad change to color chart.
Specifications
  • TestChlorine, free
  • Measurement Points0, 0.5, 1, 3, 5, 10 ppm
  • Reagent PlatformStrips
  • DescriptionFree Chlorine Test Strips, 0 to 10 ppm