LaMotte Insta-Test Alkalinity Test Strips, 0 to 180 ppm
Representative image only.
LaMotte Insta-Test Alkalinity Test Strips, 0 to 180 ppm
LaMotte | Mfr: 2997| Item: 9953262
Monitor water quality contaminants with quick spot tests
₹4,900.00INR / Pkg of 50
₹4,900.00INR / Pkg of 50
Buy It Together
Current Item
LaMotte Insta-Test Alkalinity Test Strips, 0 to 180 ppm
LaMotte Insta-Test Alkalinity Test Strips, 0 to 180 ppm
9953262
₹4,900.00INR / Pkg of 50
+
Chemetrics K-3510 Colorimetric Copper Test Kit
0029050
Call For Price
=
Total Price 4900.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Convenient, on-site water testing with bright easy-to-read colors
  • Pop-top vial with desiccant liner protects strips from moisture
  • 50 strips per vial.
More About this Item
Moisture protection, and fast, reliable results make the Insta-TEST an unbeatable system for water quality testing. Simply dip polystyrene strips into water sample, and compare the resulting color pad change to color chart.
Specifications
  • TestAlkalinity
  • Measurement Points0, 40, 80, 120, 180 ppm
  • Reagent PlatformStrips
  • DescriptionAlkalinity Test Strips, 0 to 180 ppm