Chemetrics K-7501 Colorimetric Dissolved Oxygen Test Kit , 0 to 1 ppm
Representative image only.
Chemetrics K-7501 Colorimetric Dissolved Oxygen Test Kit , 0 to 1 ppm
Chemetrics | Mfr: K-7501| Item: 0554030
Vacuum sealed reagents and handy comparator chart for quick, visual results
₹26,800.00INR / Each
₹26,800.00INR / Each
Buy It Together
Current Item
Chemetrics K-7501 Colorimetric Dissolved Oxygen Test Kit , 0 to 1 ppm
Chemetrics K-7501 Colorimetric Dissolved Oxygen Test Kit , 0 to 1 ppm
0554030
₹26,800.00INR / Each
+ + + + =
Total Price 69800.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Includes
polypropylene case with instructions; all necessary apparatus; and reagent vials for 30 tests.
More About this Item
Just break tip of ampoule in cup (included) filled with sample. Vacuum pulls sample into ampoule automatically; gently swirl to mix. Handy color comparator (included) provides accurate visual analysis. Vacuum packing keeps reagents active indefinitely.
Specifications
  • TestDissolved oxygen
  • Measurement Points0 to 1 ppm
  • Tests Per Kit30
  • DescriptionColorimetric Dissolved Oxygen Test Kit , 0 to 1 ppm