Chemetrics K-7599 Colorimetric Dissolved Oxygen Test Kit , 0 to 100 ppb
Representative image only.
Chemetrics K-7599 Colorimetric Dissolved Oxygen Test Kit , 0 to 100 ppb
Chemetrics | Mfr: K-7599| Item: 0554025
Vacuum sealed reagents and handy comparator chart for quick, visual results
₹29,300.00INR / Each
₹29,300.00INR / Each
Buy It Together
Current Item
Chemetrics K-7599 Colorimetric Dissolved Oxygen Test Kit , 0 to 100 ppb
Chemetrics K-7599 Colorimetric Dissolved Oxygen Test Kit , 0 to 100 ppb
0554025
₹29,300.00INR / Each
+
Chemetrics K-3510 Colorimetric Copper Test Kit
0029050
Call For Price
+
Chemetrics R-7540 Refill Kit for 05540-20 -25, 30/Box
0554026
₹17,600.00INR / Box of 30
+ + =
Total Price 77800.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Includes
polypropylene case with instructions; all necessary apparatus; and reagent vials for 30 tests.
More About this Item
Just break tip of ampoule in cup (included) filled with sample. Vacuum pulls sample into ampoule automatically; gently swirl to mix. Handy color comparator (included) provides accurate visual analysis. Vacuum packing keeps reagents active indefinitely.
Specifications
  • TestDissolved oxygen
  • Measurement Points0 to 100 ppb
  • Tests Per Kit30
  • DescriptionColorimetric Dissolved Oxygen Test Kit , 0 to 100 pph