Chemetrics R-2505 Refill Kit for 05540-42, 30 Tests
Representative image only.
Chemetrics R-2505 Refill Kit for 05540-42, 30 Tests
Chemetrics | Mfr: R-2505| Item: 0554043
Take it wherever you go
₹17,000.00INR / Each
₹17,000.00INR / Each
Buy It Together
Current Item
Chemetrics R-2505 Refill Kit for 05540-42, 30 Tests
Chemetrics R-2505 Refill Kit for 05540-42, 30 Tests
0554043
₹17,000.00INR / Each
+
Chemetrics K-3510 Colorimetric Copper Test Kit
0029050
Call For Price
+ + =
Total Price 61400.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Vacuum sealed packaging keeps reagents active
  • 2 year warranty
More About this Item
CHEMets colorimetric test kits let you visually compare the color of your sample to standards using a comparator, comparator sold separately. Just break tip of ampoule in cup filled with sample. Vacuum pulls sample into ampoule automatically; gently swirl to mix. Handy color comparator provides accurate visual analysis.
Two year warranty
Specifications
  • Compatible With05540-42
  • DescriptionRefill Kit for 05540-42, 30 Tests