PYREX® Brand 6402 Funnels
Squibb, separatory, pear-shaped, PYREX std taper stopper PTFE stopcock
  • Pyrex stopper, PTFE stopcock
  • No freezing or grease contamination from stopcock
  • Case quantities also available
These separatory funnels are supplied with PTFE stopcock plugs for excellent chemical resistance. The plug reduces freezing problems and grease contamination.
25,700.00 - ₹148,000.00INR / Pkg of 1
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Stopcock Plug Size Stopper Number
Availability Pricing
Pyrex 6402-60 Brand 6402 Separatory Funnel; 60 mL, pack of 1 WW-34506-01 Mfr # 6402-60
COMPARE
60 2 16
₹26,400.00INR / Pkg of 1
Pyrex 6402-125 Brand 6402 Separatory Funnel; 125 mL, pack of 1 WW-34506-02 Mfr # 6402-125
COMPARE
125 2 22
₹25,700.00INR / Pkg of 1
Pyrex 6402-250 Brand 6402 Separatory Funnel; 250 mL, pack of 1 WW-34506-03 Mfr # 6402-250
COMPARE
250 4 22
₹30,500.00INR / Pkg of 1
Pyrex 6402-500 Brand 6402 Separatory Funnel; 500 mL, pack of 1 WW-34506-04 Mfr # 6402-500
COMPARE
500 4 27
₹35,000.00INR / Pkg of 1
Pyrex 6402-125 Brand 6402 Separatory Funnel; 125 mL, case of 4 WW-34506-22 Mfr # 6402-125
COMPARE
125 2 22
₹103,000.00INR / Case of 4
Pyrex 6402-250 Brand 6402 Separatory Funnel; 250 mL, case of 4 WW-34506-23 Mfr # 6402-250
COMPARE
250 4 22
₹122,000.00INR / Case of 4
Pyrex 6402-1L Brand 6402 Separatory Funnel; 1000 mL, case of 1 WW-34506-25 Mfr # 6402-1L
COMPARE
1000 4 27
₹48,700.00INR / Case of 1
Pyrex 6402-2L Brand 6402 Separatory Funnel; 2000 mL, case of 1 WW-34506-26 Mfr # 6402-2L
COMPARE
2000 6 38
₹70,700.00INR / Case of 1
Pyrex 6402-4L Brand 6402 Separatory Funnel; 4000 mL, case of 1 WW-34506-27 Mfr # 6402-4L
COMPARE
4000 8 38
₹126,000.00INR / Case of 1
Pyrex 6402-6L Brand 6402 Separatory Funnel; 6000 mL, case of 1 WW-34506-28 Mfr # 6402-6L
COMPARE
6000 10 38
₹148,000.00INR / Case of 1
Showing 1- 10 of 10