LaMotte Sanitizer Strength Test Strips
Use these chemically treated paper strips as a quick test for sanitizer strength
  • Compare color change to included chart for easy-to-read results
Strips and color chart come supplied in a waterproof plastic vial.
2,400.00 - ₹7,000.00INR / Vial of 200
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Test Tests Per Kit Description
Availability Pricing
LaMotte 2951 Sanitizer Strength Qac Test Strips WW-99532-30 Mfr # 2951
COMPARE
QAC 100 Sanitizer Strength Qac Test Strips
₹2,400.00INR / Vial of 100
LaMotte 4250-BJ Chlorine Test Strips, Sanitizer Strength WW-99532-31 Mfr # 4250-BJ
COMPARE
Chlorine, sanitizer 200 Chlorine Test Strips, Sanitizer Strength
₹2,500.00INR / Vial of 200
LaMotte 2948-BJ Sanitizer Strength Iodine Test Strips WW-99532-32 Mfr # 2948-BJ
COMPARE
Iodine 200 Sanitizer Strength Iodine Test Strips
₹2,500.00INR / Vial of 200
LaMotte 3002 Test Strips, Chlorine Dioxide, 0-500ppm, 50 Strips/Vial 99532-34 WW-99532-34 Mfr # 3002
COMPARE
Chlorine dioxide 50 Test Strips, Chlorine Dioxide, 0-500ppm, 50 Strips/Vial 99532-34
₹4,300.00INR / Each
LaMotte 3000 Test Strips, Peracetic Acid, 0-160ppm, 50 Strips/Vial WW-99532-35 Mfr # 3000
COMPARE
Peracetic acid 50 Test Strips, Peracetic Acid, 0-160ppm, 50 Strips/Vial
₹4,000.00INR / Each
LaMotte 1910 Sanitizer Strength High Range Chlorine Test Strips WW-99532-36 Mfr # 3031
COMPARE
Chlorine, high range 50 Sanitizer Strength High Range Chlorine Test Strips
₹7,000.00INR / Pkg of 50
LaMotte 3072-J Sanitizer Strength Qac Test Strips, 0 to 500 ppm WW-99532-82 Mfr # 3072-J
COMPARE
QAC 100 Sanitizer Strength Qac Test Strips, 0 to 500 ppm
₹2,500.00INR / Pkg of 1
Showing 1- 7 of 7