LaMotte 3072-J Sanitizer Strength Qac Test Strips, 0 to 500 ppm
Representative image only.
LaMotte 3072-J Sanitizer Strength Qac Test Strips, 0 to 500 ppm
LaMotte | Mfr: 3072-J| Item: 9953282
Use these chemically treated paper strips as a quick test for sanitizer strength
₹2,700.00INR / Pkg of 1
₹2,700.00INR / Pkg of 1
Product Description
Key Features
  • Compare color change to included chart for easy-to-read results
More About this Item
Strips and color chart come supplied in a waterproof plastic vial.
Strips for QACs also include a dessiccant cap to ensure longer life.
Specifications
  • TestQAC
  • Measurement Points0, 100, 200, 300, 400, 500 ppm
  • Reagent PlatformStrips
  • Tests Per Kit100
  • DescriptionSanitizer Strength Qac Test Strips, 0 to 500 ppm