LaMotte 2951 Sanitizer Strength Qac Test Strips
Representative image only.
LaMotte 2951 Sanitizer Strength Qac Test Strips
LaMotte | Mfr: 2951| Item: 9953230
Use these chemically treated paper strips as a quick test for sanitizer strength
₹2,800.00INR / Vial of 100
₹2,800.00INR / Vial of 100
Product Description
Key Features
  • Compare color change to included chart for easy-to-read results
  • 1 year warranty
More About this Item
Strips and color chart come supplied in a waterproof plastic vial.
Strips for QACs also include a dessiccant cap to ensure longer life.
1 Year Warranty
Specifications
  • TestQAC
  • Measurement Points50, 100, 200 and 400 ppm
  • Reagent PlatformStrips
  • Tests Per Kit100
  • DescriptionSanitizer Strength Qac Test Strips