LaMotte 2948-BJ Sanitizer Strength Iodine Test Strips
Representative image only.
LaMotte 2948-BJ Sanitizer Strength Iodine Test Strips
LaMotte | Mfr: 2948-BJ| Item: 9953232
Use these chemically treated paper strips as a quick test for sanitizer strength
₹2,800.00INR / Vial of 200
₹2,800.00INR / Vial of 200
Product Description
Key Features
  • Compare color change to included chart for easy-to-read results
  • 1 year warranty
More About this Item
Strips and color chart come supplied in a waterproof plastic vial.
1 Year Warranty
Specifications
  • TestIodine
  • Measurement Points12, 25, 50, and 100 ppm
  • Reagent PlatformStrips
  • Tests Per Kit200
  • DescriptionSanitizer Strength Iodine Test Strips