LaMotte 1910 Sanitizer Strength High Range Chlorine Test Strips
Representative image only.
LaMotte 1910 Sanitizer Strength High Range Chlorine Test Strips
LaMotte | Mfr: 3031| Item: 9953236
Use these chemically treated paper strips as a quick test for sanitizer strength
₹6,300.00INR / Pkg of 50
₹6,300.00INR / Pkg of 50
Product Description
Key Features
  • Compare color change to included chart for easy-to-read results
More About this Item
Strips and color chart come supplied in a waterproof plastic vial.
Specifications
  • TestChlorine, high range
  • Measurement Points0, 50, 100, 250, 500, and 800 ppm
  • Reagent PlatformStrips
  • Tests Per Kit50
  • DescriptionSanitizer Strength High Range Chlorine Test Strips