LaMotte 4250-BJ Chlorine Test Strips, Sanitizer Strength
Representative image only.
LaMotte 4250-BJ Chlorine Test Strips, Sanitizer Strength
LaMotte | Mfr: 4250-BJ| Item: 9953231
Use these chemically treated paper strips as a quick test for sanitizer strength
₹2,900.00INR / Vial of 200
₹2,900.00INR / Vial of 200
Product Description
Key Features
  • Compare color change to included chart for easy-to-read results
  • 1 year warranty
More About this Item
Strips and color chart come supplied in a waterproof plastic vial.
1 Year Warranty
Specifications
  • TestChlorine, sanitizer
  • Measurement Points10, 50, 100, and 200 ppm
  • Reagent PlatformStrips
  • Tests Per Kit200
  • DescriptionChlorine Test Strips, Sanitizer Strength