LaMotte 3000 Test Strips, Peracetic Acid, 0-160ppm, 50 Strips/Vial
Representative image only.
LaMotte 3000 Test Strips, Peracetic Acid, 0-160ppm, 50 Strips/Vial
LaMotte | Mfr: 3000| Item: 9953235
Use these chemically treated paper strips as a quick test for sanitizer strength
₹4,000.00INR / Each
₹4,000.00INR / Each
Product Description
Key Features
  • Compare color change to included chart for easy-to-read results
  • 1 year warranty
More About this Item
Strips and color chart come supplied in a waterproof plastic vial.
Test strips allow to easily determine peracetic acid levels levels from 0-160ppm. Simply swirl a strip in your sample and 10 seconds later read it.
1 Year Warranty
Specifications
  • TestPeracetic acid
  • Reagent PlatformStrips
  • Tests Per Kit50
  • DescriptionTest Strips, Peracetic Acid, 0-160ppm, 50 Strips/Vial