Spectra Por S/P 1 Dialysis Membrane Trial Kit, 6000-8000 Dalton, 50mm Flat Width

Representative image only.

Spectra Por S/P 1 Dialysis Membrane Trial Kit, 6000-8000 Dalton, 50mm Flat Width

Spectra Por | Mfr: 132665T| Item: 0290001

Contains everything you needed to evaluate dialysis membranes

₹41,700.00INR / Each

₹41,700.00INR / Each

Buy It Together

Current Item

Spectra Por S/P 1 Dialysis Membrane Trial Kit, 6000-8000 Dalton, 50mm Flat Width
Spectra Por S/P 1 Dialysis Membrane Trial Kit, 6000-8000 Dalton, 50mm Flat Width
0290001

₹41,700.00INR / Each
+ + + + + + + =
Total Price 490900.00 ADD 8 ITEMS TO CART

Customers Also Bought

Product Description

Includes

A 5-m roll of Spectra/Por dialysis tubing, one standard Spectra/Por closure, and one weighted Spectra/Por closure.

Key Features

  • Ideal for smaller dialysis experiments
  • 1 year warranty

More About this Item

These convenient kits contain everything you need for smaller dialysis experiments or evaluating dialysis tubing for a dialysis application. The small roll is ideal for low volume projects. Spectra/Por® 1 through 3 Dialysis Trail Kits are available in a range of nominal molecular weight cut-offs (MWCO) for less specific separations. Spectra/Por grades 1 through 3 are packaged dry with 10% glycerol and contain trace levels of heavy metals and sulfur.

1 Year Warranty
Specifications
  • MaterialRegenerated Cellulose
  • Max Molecular Weight Cut Off (Mwco) (Dalton)8000
  • Min Molecular Weight Cut Off (Mwco) (Dalton)6000
  • Flat Width (mm)50
  • Diameter (mm)32
  • Length Per Pack (ft)16.5
  • Diameter (in)1.26
  • DescriptionDialysis Membrane Trial Kit, 6000-8000Dalton 50mm
  • Warranty1 year
Reviews
Q&A