Spectra/Por 6 Prewetted Regenerated Cellulose (RC) Tubing
Available in a variety of defined MWCO's for specific retentivity
  • Made of regenerated cellulose (RC) for gentle separation and purification of solutes.
Prewetted in 0.05% sodium azide solution for easier use; contains traces of sulfur and heavy metals. Packaged in 50-ft lengths.
98,800.00 - ₹116,000.00INR / Box of 1
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Flat Width (mm) Max Molecular Weight Cut Off (Mwco) (Dalton)
Availability Pricing
Spectra Por S/P 6 6 RC Dialysis Tubing, 2000 MWCO, 18mm/11.5mm; 50ft 0289031 Mfr # 132620
COMPARE
11.5 18 2000
₹102,000.00INR / Box of 1
Spectra Por S/P 6 6 RC Dialysis Tubing, 2000 MWCO, 38mm/24mm; 50ft 0289032 Mfr # 132625
COMPARE
24 38 2000
₹102,000.00INR / Box of 1
Spectra Por S/P 6 6 RC Dialysis Tubing, 2000 MWCO, 45mm/29mm; 50ft 0289033 Mfr # 132633
COMPARE
29 45 2000
₹102,000.00INR / Box of 1
Spectra Por S/P 6 6 RC Dialysis Tubing, 10000 MWCO, 45mm/29mm; 50ft 0289038 Mfr # 132576
COMPARE
29 45 10000
₹102,000.00INR / Box of 1
Spectra Por S/P 6 6 RC Dialysis Tubing, 15000 MWCO, 32mm/20.4mm; 50ft 0289041 Mfr # 132564
COMPARE
20.4 32 15000
₹102,000.00INR / Box of 1
Spectra Por S/P 6 6 RC Dialysis Tubing, 15000 MWCO, 45mm/29mm; 50ft 0289042 Mfr # 132566
COMPARE
29 45 15000
₹102,000.00INR / Box of 1
Spectra Por S/P 6 6 Membranes; MWCO, 1000; flat width, 18mm; diameter, 11.5mm 0290121 Mfr # 132636
COMPARE
11.5 18 1000
₹98,800.00INR / Pkg of 1
Spectra Por S/P 6 6 Membranes; MWCO, 1000; flat width, 38mm; diameter, 24.2mm 0290122 Mfr # 132638
COMPARE
24.2 38 1000
₹98,800.00INR / Pkg of 1
Spectra Por S/P 6 6 Membranes; MWCO, 1000; flat width, 45mm; diameter, 28.6mm 0290123 Mfr # 132640
COMPARE
28.6 45 1000
₹116,000.00INR / Pkg of 1
Spectra Por S/P 6 6 Membranes; MWCO, 3500; flat width, 45mm; diameter, 28.6mm 0290131 Mfr # 132592
COMPARE
28.6 45 3500
₹116,000.00INR / Pkg of 1
Spectra Por S/P 6 6 Membranes; MWCO, 8000; flat width, 40mm; diameter, 25.5mm 0290142 Mfr # 132584
COMPARE
25.5 40 8000
₹98,800.00INR / Pkg of 1
Spectra Por S/P 6 6 Membranes; MWCO, 25,000; flat width, 28mm; diameter, 17.8mm 0290171 Mfr # 132552
COMPARE
17.8 28 25000
₹98,800.00INR / Pkg of 1
Spectra Por S/P 6 6 Membranes; MWCO, 25,000; flat width, 34mm; diameter, 21.6mm 0290172 Mfr # 132554
COMPARE
21.6 34 25000
₹98,800.00INR / Pkg of 1
Spectra Por S/P 6 6 Membranes; MWCO, 50,000; flat width, 12mm; diameter, 7.6mm 0290180 Mfr # 132540
COMPARE
7.6 12 50000
₹98,800.00INR / Pkg of 1
Spectra Por S/P 6 6 Membranes; MWCO, 50,000; flat width, 28mm; diameter, 17.8mm 0290181 Mfr # 132542
COMPARE
17.8 28 50000
₹98,800.00INR / Pkg of 1
Spectra Por S/P 6 6 Membranes; MWCO, 50,000; flat width, 34mm; length, 21.6mm 0290182 Mfr # 132544
COMPARE
21.6 34 50000
₹98,800.00INR / Pkg of 1
Showing 1- 16 of 16