Spectra Por S/P 6 6 RC Dialysis Tubing, 15000 MWCO, 32mm/20.4mm; 50ft

Representative image only.

Spectra Por S/P 6 6 RC Dialysis Tubing, 15000 MWCO, 32mm/20.4mm; 50ft

Spectra Por | Mfr: 132564| Item: 0289041

Available in a variety of defined MWCO's for specific retentivity

₹102,000.00INR / Box of 1

₹102,000.00INR / Box of 1

Buy It Together

Current Item

Spectra Por S/P 6 6 RC Dialysis Tubing, 15000 MWCO, 32mm/20.4mm; 50ft
Spectra Por S/P 6 6 RC Dialysis Tubing, 15000 MWCO, 32mm/20.4mm; 50ft
0289041

₹102,000.00INR / Box of 1
+ =
Total Price 134700.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Made of regenerated cellulose (RC) for gentle separation and purification of solutes.
  • 1 year warranty

More About this Item

Prewetted in 0.05% sodium azide solution for easier use; contains traces of sulfur and heavy metals. Packaged in 50-ft lengths.

1 Year Warranty
Specifications
  • MaterialRegenerated Cellulose
  • Max Molecular Weight Cut Off (Mwco) (Dalton)15000
  • Flat Width (mm)32
  • Diameter (mm)20.4
  • Length Per Pack (ft)16
  • Diameter (in)0.803
  • Description6 RC Dialysis Tubing, 15000 MWCO, 32mm/20.4mm; 50ft
  • Warranty1 year
Reviews
Q&A