Spectra Por S/P 6 6 Membranes; MWCO, 1000; flat width, 18mm; diameter, 11.5mm

Representative image only.

Spectra Por S/P 6 6 Membranes; MWCO, 1000; flat width, 18mm; diameter, 11.5mm

Spectra Por | Mfr: 132636| Item: 0290121

Available in a variety of defined MWCO's for specific retentivity

₹98,800.00INR / Pkg of 1

₹98,800.00INR / Pkg of 1

Buy It Together

Current Item

Spectra Por S/P 6 6 Membranes; MWCO, 1000; flat width, 18mm; diameter, 11.5mm
Spectra Por S/P 6 6 Membranes; MWCO, 1000; flat width, 18mm; diameter, 11.5mm
0290121

₹98,800.00INR / Pkg of 1
+ =
Total Price 131500.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Made of regenerated cellulose (RC) for gentle separation and purification of solutes.
  • 6 month warranty

More About this Item

Prewetted in 0.05% sodium azide solution for easier use; contains traces of sulfur and heavy metals. Packaged in 50-ft lengths.

6 Month Warranty
Specifications
  • MaterialRegenerated Cellulose
  • Max Molecular Weight Cut Off (Mwco) (Dalton)1000
  • Flat Width (mm)18
  • Diameter (mm)11.5
  • Length Per Pack (ft)16
  • Diameter (in)0.453
  • Description6 Membranes; MWCO, 1000; flat width, 18mm; diameter, 11.5mm
  • Warranty6 month
Reviews
Q&A