Universal dialysis tubing closure for all membrane types
The most convenient method for sealing dialysis tubing without the risk of leaking
  • Easy to open and reseal
Available in several colors for easy sample identification. Select closures with a sealing width 4 to 10 mm longer than the flat width of the dialysis tubing.
24,400.00 - ₹38,100.00INR / Box of 10
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
Compareitem specifications
Color Tubing Sealing Width (mm)
Availability Pricing
Spectra Por 142152 Universal Dialysis Tubing Closure, 50 mm, Red; 10/Bx 0289970 Mfr # 142152
COMPARE
Red 50
₹24,400.00INR / Box of 10
Spectra Por 142150 Dialysis Tubing Closure, White; max sealing width 50 mm; pack of 10. 0289971 Mfr # 142150
COMPARE
White 50
₹24,400.00INR / Pkg of 10
Spectra Por 142153 Dialysis Tubing Closure, Yellow; max sealing width 50 mm; pack of 10. 0289972 Mfr # 142153
COMPARE
Yellow 50
₹24,400.00INR / Pkg of 10
Spectra Por 142154 Dialysis Tubing Closure, Blue; max sealing width 50 mm; pack of 10. 0289973 Mfr # 142154
COMPARE
Blue 50
₹28,400.00INR / Pkg of 10
Spectra Por 142170 Dialysis Tubing Closure, White; max sealing width 70 mm; pack of 10. 0289974 Mfr # 142170
COMPARE
White 70
₹27,400.00INR / Pkg of 10
Spectra Por 142172 Dialysis Tubing Closure, Red; max sealing width 70 mm; pack of 10. 0289975 Mfr # 142172
COMPARE
Red 70
₹31,900.00INR / Pkg of 10
Spectra Por 142173 Dialysis Tubing Closure, Yellow; max sealing width 70 mm; pack of 10. 0289976 Mfr # 142173
COMPARE
Yellow 70
₹27,400.00INR / Pkg of 10
Spectra Por 142174 Dialysis Tubing Closure, Blue; max sealing width 70 mm; pack of 10. 0289977 Mfr # 142174
COMPARE
Blue 70
₹27,400.00INR / Pkg of 10
Spectra Por 142110 Dialysis Tubing Closure, White; max sealing width 110 mm; pack of 10. 0289978 Mfr # 142110
COMPARE
White 110
₹30,000.00INR / Pkg of 10
Spectra Por 142112 Dialysis Tubing Closure, Red; max sealing width 110 mm; pack of 10. 0289979 Mfr # 142112
COMPARE
Red 110
₹30,000.00INR / Pkg of 10
Spectra Por 142113 Dialysis Tubing Closure, Yellow; max sealing width 110 mm; pack of 10. 0289980 Mfr # 142113
COMPARE
Yellow 110
₹30,000.00INR / Pkg of 10
Spectra Por 142250 Dialysis Tubing Closure, White 150MM; 10/pk 0289991 Mfr # 142250
COMPARE
White 150
₹32,700.00INR / Pkg of 10
Spectra Por 142252 Dialysis Tubing Closure, Red 150MM; 10/pk 0289992 Mfr # 142252
COMPARE
Red 150
₹38,100.00INR / Pkg of 10
Spectra Por 142253 Dialysis Tubing Closure, Yellow 150MM; 10/pk 0289993 Mfr # 142253
COMPARE
Yellow 150
₹32,700.00INR / Pkg of 10
Showing 1- 14 of 14