Spectra Por 142173 Dialysis Tubing Closure, Yellow; max sealing width 70 mm; pack of 10.
Representative image only.
Spectra Por 142173 Dialysis Tubing Closure, Yellow; max sealing width 70 mm; pack of 10.
Spectra Por | Mfr: 142173| Item: 0289976
The most convenient method for sealing dialysis tubing without the risk of leaking
₹27,400.00INR / Pkg of 10
₹27,400.00INR / Pkg of 10
Buy It Together
Current Item
Spectra Por 142173 Dialysis Tubing Closure, Yellow; max sealing width 70 mm; pack of 10.
Spectra Por 142173 Dialysis Tubing Closure, Yellow; max sealing width 70 mm; pack of 10.
0289976
₹27,400.00INR / Pkg of 10
+ =
Total Price 63100.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Easy to open and reseal
  • 1 year warranty
More About this Item
Available in several colors for easy sample identification. Select closures with a sealing width 4 to 10 mm longer than the flat width of the dialysis tubing.
1 Year Warranty
Specifications
  • Tubing Sealing Width (mm)70
  • ColorYellow
  • DescriptionDialysis Tubing Closure, Yellow; max sealing width 70 mm; pack of 10.