Spectra Por 142112 Dialysis Tubing Closure, Red; max sealing width 110 mm; pack of 10.
Representative image only.
Spectra Por 142112 Dialysis Tubing Closure, Red; max sealing width 110 mm; pack of 10.
Spectra Por | Mfr: 142112| Item: 0289979
The most convenient method for sealing dialysis tubing without the risk of leaking
₹30,000.00INR / Pkg of 10
₹30,000.00INR / Pkg of 10
Buy It Together
Current Item
Spectra Por 142112 Dialysis Tubing Closure, Red; max sealing width 110 mm; pack of 10.
Spectra Por 142112 Dialysis Tubing Closure, Red; max sealing width 110 mm; pack of 10.
0289979
₹30,000.00INR / Pkg of 10
+ =
Total Price 65700.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Easy to open and reseal
  • 1 year warranty
More About this Item
Available in several colors for easy sample identification. Select closures with a sealing width 4 to 10 mm longer than the flat width of the dialysis tubing.
1 Year Warranty
Specifications
  • Tubing Sealing Width (mm)110
  • ColorRed
  • DescriptionDialysis Tubing Closure, Red; max sealing width 110 mm; pack of 10.