Spectra/Por Biotech-Grade Dialysis Tubing Trial Kits
These convenient kits contain everythig needed to evaluate dialysis membranes for your application. The small roll is also ideal for low volume projects.
Cellulose Ester (CE) offers a wider range of more precise MWCO’s for critical separations; packaged wet in 0.05% sodium azide.
38,400.00 - ₹44,800.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Material Description
Availability Pricing
Spectra Por 131054T Biotech-Grade Dialysis Tubing Trial Kit, 0.1-0.5 kDalton, 16 mm 0298011 Mfr # 131054T
COMPARE
10 Cellulose Ester Biotech-Grade Dialysis Tubing Trial Kit, 0.1-0.5 kDalton, 16 mm
₹44,800.00INR / Each
Spectra Por 131090T Biotech-Grade Dialysis Tubing Trial Kit, 0.5-1.0 kDalton, 16 mm 0298012 Mfr # 131090T
COMPARE
10 Cellulose Ester Biotech-Grade Dialysis Tubing Trial Kit, 0.5-1.0 kDalton, 16 mm
₹44,800.00INR / Each
Spectra Por 131198T Biotech-Grade Dialysis Tubing Trial Kit, 3.5-5.0 kDalton, 16 mm 0298013 Mfr # 131198T
COMPARE
10 Cellulose Ester Biotech-Grade Dialysis Tubing Trial Kit, 3.5-5.0 kDalton, 16 mm
₹44,800.00INR / Each
Spectra Por 131270T Biotech-Grade Dialysis Tubing Trial Kit, 8.0-10.0 kDalton, 16 mm 0298014 Mfr # 131270T
COMPARE
10 Cellulose Ester Biotech-Grade Dialysis Tubing Trial Kit, 8.0-10.0 kDalton, 16 mm
₹38,400.00INR / Each
Spectra Por 131378T Biotech-Grade Dialysis Tubing Trial Kit, 50 kDalton, 16 mm 0298016 Mfr # 131378T
COMPARE
10 Cellulose Ester Biotech-Grade Dialysis Tubing Trial Kit, 50 kDalton, 16 mm
₹38,400.00INR / Each
Spectra Por 131414T Biotech-Grade Dialysis Tubing Trial Kit, 100 kDalton, 16 mm 0298017 Mfr # 131414T
COMPARE
10 Cellulose Ester Biotech-Grade Dialysis Tubing Trial Kit, 100 kDalton, 16 mm
₹38,400.00INR / Each
Spectra Por 131450T Biotech-Grade Dialysis Tubing Trial Kit, 300 kDalton, 16 mm 0298018 Mfr # 131450T
COMPARE
10 Cellulose Ester Biotech-Grade Dialysis Tubing Trial Kit, 300 kDalton, 16 mm
₹38,400.00INR / Each
Spectra Por 131486T Biotech-Grade Dialysis Tubing Trial Kit, 1,000 kDalton, 16 mm 0298019 Mfr # 131486T
COMPARE
10 Cellulose Ester Biotech-Grade Dialysis Tubing Trial Kit, 1,000 kDalton, 16 mm
₹38,400.00INR / Each
Showing 1- 8 of 8