Spectra Por 131450T Biotech-Grade Dialysis Tubing Trial Kit, 300 kDalton, 16 mm
Representative image only.
Spectra Por 131450T Biotech-Grade Dialysis Tubing Trial Kit, 300 kDalton, 16 mm
Spectra Por | Mfr: 131450T| Item: 0298018
₹38,400.00INR / Each
₹38,400.00INR / Each
Buy It Together
Current Item
Spectra Por 131450T Biotech-Grade Dialysis Tubing Trial Kit, 300 kDalton, 16 mm
Spectra Por 131450T Biotech-Grade Dialysis Tubing Trial Kit, 300 kDalton, 16 mm
0298018
₹38,400.00INR / Each
+ + =
Total Price 131300.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Includes
1 m roll of Biotech CE dialysis tubing, 2 universal closures, and 5 opening picks
Key Features
  • 1 year warranty
More About this Item
These convenient kits contain everythig needed to evaluate dialysis membranes for your application. The small roll is also ideal for low volume projects.
Cellulose Ester (CE) offers a wider range of more precise MWCO’s for critical separations; packaged wet in 0.05% sodium azide.
1 Year Warranty
Specifications
  • MaterialCellulose Ester
  • Diameter (mm)10
  • DescriptionBiotech-Grade Dialysis Tubing Trial Kit, 300 kDalton, 16 mm